missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Diethoxyethane 98.0+%, TCI America™
$42.87 - $219.95
Chemical Identifiers
| CAS | 629-14-1 |
|---|---|
| Molecular Formula | C6H14O2 |
| Molecular Weight (g/mol) | 118.176 |
| MDL Number | MFCD00009253 |
| InChI Key | LZDKZFUFMNSQCJ-UHFFFAOYSA-N |
| Synonym | ethylene glycol diethyl ether, diethyl cellosolve, ethane, 1,2-diethoxy, hisolve eme, 3,6-dioxaoctane, glyme-1, diethyl glycol, 2-ethoxyethyl ethyl ether, egdee, diethylether ethylenglykolu |
| PubChem CID | 12375 |
| IUPAC Name | 1,2-diethoxyethane |
| SMILES | CCOCCOCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D045625ML
|
TCI America
D045625ML |
25 mL |
Each for $42.87
|
|
|||||
|
D0456500ML
|
TCI America
D0456500ML |
500 mL |
Each for $219.95
|
|
|||||
Chemical Identifiers
| 629-14-1 | |
| 118.176 | |
| LZDKZFUFMNSQCJ-UHFFFAOYSA-N | |
| 12375 | |
| CCOCCOCC |
| C6H14O2 | |
| MFCD00009253 | |
| ethylene glycol diethyl ether, diethyl cellosolve, ethane, 1,2-diethoxy, hisolve eme, 3,6-dioxaoctane, glyme-1, diethyl glycol, 2-ethoxyethyl ethyl ether, egdee, diethylether ethylenglykolu | |
| 1,2-diethoxyethane |
Specifications
| 629-14-1 | |
| 121°C | |
| MFCD00009253 | |
| 1153 | |
| LZDKZFUFMNSQCJ-UHFFFAOYSA-N | |
| 1,2-diethoxyethane | |
| 12375 | |
| ≥98.0% (GC) | |
| 1,2-Diethoxyethane |
| Colorless | |
| C6H14O2 | |
| 25 mL | |
| ethylene glycol diethyl ether, diethyl cellosolve, ethane, 1,2-diethoxy, hisolve eme, 3,6-dioxaoctane, glyme-1, diethyl glycol, 2-ethoxyethyl ethyl ether, egdee, diethylether ethylenglykolu | |
| CCOCCOCC | |
| 118.176 | |
| 118.18 | |
| Liquid |
Safety and Handling
EINECSNumber : (7)-1321
RTECSNumber : KI1225000
TSCA : Yes