missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diethoxyethane 98.0+%, TCI America™

Catalog No. D0456500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0456500ML 500 mL
D045625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0456500ML Supplier TCI America Supplier No. D0456500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 629-14-1
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.176
MDL Number MFCD00009253
InChI Key LZDKZFUFMNSQCJ-UHFFFAOYSA-N
Synonym ethylene glycol diethyl ether, diethyl cellosolve, ethane, 1,2-diethoxy, hisolve eme, 3,6-dioxaoctane, glyme-1, diethyl glycol, 2-ethoxyethyl ethyl ether, egdee, diethylether ethylenglykolu
PubChem CID 12375
IUPAC Name 1,2-diethoxyethane
SMILES CCOCCOCC

Specifications

Color Colorless
Boiling Point 121°C
Quantity 500 mL
UN Number 1153
Formula Weight 118.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Diethoxyethane
EINECSNumber (7)-1321
RTECSNumber KI1225000
TSCA Yes