missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diethoxyethane, 98%

Catalog No. AAL14283AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL14283AE 100 mL
AAL14283AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL14283AE Supplier Thermo Scientific Chemicals Supplier No. L14283AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 629-14-1 | C6H14O2 | 118.176 g/mol

Ethylene glycol diethyl ether is a important solvent, used for ink, paint and coating industry. Mainly used in polymer, electrochemistry, boracium chemistry, resin, nitro cellulose, In this product , the benzene, xenene, terphenyl, anthracene reacts with sodium and then bring about the complex; it is also used in surface treatment, halogen analysis in gasoline, acetic acid recycle in diluted acetic acid, depaint agent, thinner, flushing agent, stabilizer, anti oxidant , thickener of lube.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylene glycol diethyl ether is a important solvent, used for ink, paint and coating industry. Mainly used in polymer, electrochemistry, boracium chemistry, resin, nitro cellulose, In this product , the benzene, xenene, terphenyl, anthracene reacts with sodium and then bring about the complex; it is also used in surface treatment, halogen analysis in gasoline, acetic acid recycle in diluted acetic acid, depaint agent, thinner, flushing agent, stabilizer, anti oxidant , thickener of lube.

Solubility
Soluble in water 34 g/L @ 20˚C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.

Chemical Identifiers

CAS 629-14-1
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.176
MDL Number MFCD00009253
InChI Key LZDKZFUFMNSQCJ-UHFFFAOYSA-N
Synonym ethylene glycol diethyl ether, diethyl cellosolve, ethane, 1,2-diethoxy, hisolve eme, 3,6-dioxaoctane, glyme-1, diethyl glycol, 2-ethoxyethyl ethyl ether, egdee, diethylether ethylenglykolu
PubChem CID 12375
IUPAC Name 1,2-diethoxyethane
SMILES CCOCCOCC

Specifications

Melting Point -74°C
Density 0.842
Boiling Point 120°C to 121°C
Flash Point 19°C (66°F)
Refractive Index 1.392
Linear Formula CH3CH2OCH2CH2OCH2CH3
Quantity 100 mL
UN Number UN1153
Beilstein 1732917
Solubility Information Soluble in water 34g/L @ 20?C.
Formula Weight 118.18
Percent Purity 98%
Chemical Name or Material 1,2-Diethoxyethane
Show More Show Less
Hazard Category H225-H319-H360Df
Hazard Statement GHS H Statement
H225-H360-H319
Highly flammable liquid and vapor.
May damage fertility or the unborn child.
Causes serious eye irritation.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P264b-P280i-P281-P303+P361+P353-P305+P351+P338-P308+P313-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYLENE GLYCOL DIETHYL ETHER
EINECSNumber 211-076-1
RTECSNumber KI1225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only