missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dichloropropane 98.0+%, TCI America™
$44.28 - $192.52
Chemical Identifiers
| CAS | 78-87-5 |
|---|---|
| Molecular Formula | C3H6Cl2 |
| Molecular Weight (g/mol) | 112.98 |
| MDL Number | MFCD00000868 |
| InChI Key | KNKRKFALVUDBJE-UHFFFAOYNA-N |
| Synonym | propylene dichloride, propane, 1,2-dichloro, propylene chloride, dwuchloropropan, bichlorure de propylene, rcra waste number u083, caswell no. 324, dwuchloropropan polish, alpha,beta-dichloropropane, ccris 951 |
| PubChem CID | 6564 |
| ChEBI | CHEBI:82163 |
| IUPAC Name | 1,2-dichloropropane |
| SMILES | CC(Cl)CCl |
Chemical Identifiers
| 78-87-5 | |
| 112.98 | |
| KNKRKFALVUDBJE-UHFFFAOYNA-N | |
| 6564 | |
| 1,2-dichloropropane |
| C3H6Cl2 | |
| MFCD00000868 | |
| propylene dichloride, propane, 1,2-dichloro, propylene chloride, dwuchloropropan, bichlorure de propylene, rcra waste number u083, caswell no. 324, dwuchloropropan polish, alpha,beta-dichloropropane, ccris 951 | |
| CHEBI:82163 | |
| CC(Cl)CCl |
Specifications
| 78-87-5 | |
| 97°C | |
| MFCD00000868 | |
| 1279 | |
| KNKRKFALVUDBJE-UHFFFAOYNA-N | |
| 1,2-dichloropropane | |
| 6564 | |
| 112.98 | |
| Liquid |
| Yellow | |
| C3H6Cl2 | |
| 25 g | |
| propylene dichloride, propane, 1,2-dichloro, propylene chloride, dwuchloropropan, bichlorure de propylene, rcra waste number u083, caswell no. 324, dwuchloropropan polish, alpha,beta-dichloropropane, ccris 951 | |
| CC(Cl)CCl | |
| 112.98 | |
| CHEBI:82163 | |
| ≥98.0% (GC) | |
| 1,2-Dichloropropane |
Safety and Handling
EINECSNumber : (2)-0081
RTECSNumber : TX9625000
TSCA : Yes