missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloropropane 98.0+%, TCI America™

Catalog No. D0398500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0398500G 500 g
D039825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0398500G Supplier TCI America Supplier No. D0398500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 78-87-5
Molecular Formula C3H6Cl2
Molecular Weight (g/mol) 112.98
MDL Number MFCD00000868
InChI Key KNKRKFALVUDBJE-UHFFFAOYNA-N
Synonym propylene dichloride, propane, 1,2-dichloro, propylene chloride, dwuchloropropan, bichlorure de propylene, rcra waste number u083, caswell no. 324, dwuchloropropan polish, alpha,beta-dichloropropane, ccris 951
PubChem CID 6564
ChEBI CHEBI:82163
IUPAC Name 1,2-dichloropropane
SMILES CC(Cl)CCl

Specifications

Color Yellow
Boiling Point 97°C
Quantity 500 g
UN Number 1279
Formula Weight 112.98
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Dichloropropane
EINECSNumber (2)-0081
RTECSNumber TX9625000
TSCA Yes