missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloropropane, 98%

CAS: 78-87-5 | C3H6Cl2 | 112.98 g/mol

$45.90 - $122.68

Chemical Identifiers

CAS 78-87-5
Molecular Formula C3H6Cl2
Molecular Weight (g/mol) 112.98
MDL Number MFCD00000868
InChI Key KNKRKFALVUDBJE-UHFFFAOYNA-N
Synonym propylene dichloride, propane, 1,2-dichloro, propylene chloride, dwuchloropropan, bichlorure de propylene, rcra waste number u083, caswell no. 324, dwuchloropropan polish, alpha,beta-dichloropropane, ccris 951
PubChem CID 6564
ChEBI CHEBI:82163
IUPAC Name 1,2-dichloropropane
SMILES CC(Cl)CCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0308314
View Documents
Thermo Scientific Chemicals
L0308314
25 g
Each for $45.90
Only null left
 
AAL0308322
View Documents
Thermo Scientific Chemicals
L0308322
100 g
Each for $122.68
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

78-87-5
112.98
KNKRKFALVUDBJE-UHFFFAOYNA-N
6564
1,2-dichloropropane
C3H6Cl2
MFCD00000868
propylene dichloride, propane, 1,2-dichloro, propylene chloride, dwuchloropropan, bichlorure de propylene, rcra waste number u083, caswell no. 324, dwuchloropropan polish, alpha,beta-dichloropropane, ccris 951
CHEBI:82163
CC(Cl)CCl

Specifications

78-87-5
1.157
4°C (39°F)
C3H6Cl2
MFCD00000868
UN1279
14,7854
KNKRKFALVUDBJE-UHFFFAOYNA-N
1,2-dichloropropane
6564
112.99
1,2-Dichloropropane
-100°C
95°C to 97°C
Sweet
1.438
25 g
1718880
propylene dichloride, propane, 1,2-dichloro, propylene chloride, dwuchloropropan, bichlorure de propylene, rcra waste number u083, caswell no. 324, dwuchloropropan polish, alpha,beta-dichloropropane, ccris 951
CC(Cl)CCl
112.98
CHEBI:82163
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H302-H332
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.

P201-P202-P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P308+P313-P330-P370+P378q-P501c

H225-H302+H332-H350

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 1,2-DICHLOROPROPANE

EINECSNumber : 201-152-2

RTECSNumber : TX9625000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only