missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloropropane, 98%

Catalog No. AAL0308322
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0308322 100 g
AAL0308314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0308322 Supplier Thermo Scientific Chemicals Supplier No. L0308322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 78-87-5 | C3H6Cl2 | 112.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78-87-5
Molecular Formula C3H6Cl2
Molecular Weight (g/mol) 112.98
MDL Number MFCD00000868
InChI Key KNKRKFALVUDBJE-UHFFFAOYNA-N
Synonym propylene dichloride, propane, 1,2-dichloro, propylene chloride, dwuchloropropan, bichlorure de propylene, rcra waste number u083, caswell no. 324, dwuchloropropan polish, alpha,beta-dichloropropane, ccris 951
PubChem CID 6564
ChEBI CHEBI:82163
IUPAC Name 1,2-dichloropropane
SMILES CC(Cl)CCl

Specifications

Melting Point -100°C
Density 1.157
Boiling Point 95°C to 97°C
Flash Point 4°C (39°F)
Odor Sweet
Refractive Index 1.438
Quantity 100 g
UN Number UN1279
Beilstein 1718880
Merck Index 14,7854
Formula Weight 112.99
Percent Purity 98%
Chemical Name or Material 1,2-Dichloropropane
Show More Show Less
Hazard Category H225-H302+H332-H350
Hazard Statement GHS H Statement
H225-H302-H332
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P308+P313-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 1,2-DICHLOROPROPANE
EINECSNumber 201-152-2
RTECSNumber TX9625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only