missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloropropane, 98%, pure

Catalog No. AC113670500
Change view
Click to view available options
Quantity:
5 mL
50 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC113670500 50 mL Glass bottle
AC113670050 5 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC113670500 Supplier Thermo Scientific Chemicals Supplier No. 113670500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 78-87-5 | C3H6Cl2 | 112.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78-87-5
Molecular Formula C3H6Cl2
Molecular Weight (g/mol) 112.98
MDL Number MFCD00000868
InChI Key KNKRKFALVUDBJE-UHFFFAOYNA-N
Synonym propylene dichloride, propane, 1,2-dichloro, propylene chloride, dwuchloropropan, bichlorure de propylene, rcra waste number u083, caswell no. 324, dwuchloropropan polish, alpha,beta-dichloropropane, ccris 951
PubChem CID 6564
ChEBI CHEBI:82163
IUPAC Name 1,2-dichloropropane
SMILES CC(Cl)CCl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -100.0°C
Color Colorless to Yellow
Density 1.1560g/mL
Boiling Point 95.0°C to 96.0°C
Flash Point 15°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4380 to 1.4400
Linear Formula CH3CH(Cl)CH2Cl
Quantity 50 mL
Beilstein 01, 105
Specific Gravity 1.156
Merck Index 15, 7967
Solubility Information Solubility in water: 3g/L (20°C). Other solubilities: soluble in most common organic solvents
Formula Weight 112.99
Percent Purity 98%
Grade Pure
Physical Form Liquid
Chemical Name or Material 1, 2-Dichloropropane, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapour.
Harmful if swallowed.
Harmful if inhaled.
May cause cancer.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi
EINECSNumber 201-152-2
RTECSNumber TX9625000
TSCA TSCA

RUO – Research Use Only