missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloroethane, Spectrophotometric Grade, 99+%

Catalog No. p-7049975
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA32462K2 1 L
AA32462K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA32462K2 Supplier Thermo Scientific Chemicals Supplier No. 032462K2
Only null left

CAS: 107-06-2 | C2H4Cl2 | 98.95 g/mol

1,2-dichloroethane is used in the production of vinyl chloride which is used to make a variety of plastic and vinyl products including polyvinyl chloride (PVC) pipes, furniture and automobile upholstery, wall coverings, house wares, and automobile parts.1,2-Dichloroethane is also used generally as an intermediate for other organic chemical compounds and as a solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 107-06-2
Molecular Formula C2H4Cl2
Molecular Weight (g/mol) 98.95
MDL Number MFCD00000963
InChI Key WSLDOOZREJYCGB-UHFFFAOYSA-N
Synonym ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion
PubChem CID 11
ChEBI CHEBI:27789
IUPAC Name 1,2-dichloroethane
SMILES ClCCCl

Specifications

Melting Point -35°C
Density 1.256 g/mL
Quantity 1 L
Flash Point 15°C (59°F)
Physical Form Liquid
Refractive Index 1.4448
UN Number UN1184
Beilstein 605264
Merck Index 14,3797
Boiling Point 83°C
Solubility Information Soluble in water 0.87g/100 mL (20°C).
Formula Weight 98.96
Percent Purity ≥99%
Grade Spectrophotometry
Chemical Name or Material 1,2-Dichloroethane
Show More Show Less
Hazard Category H225-H302-H315-H319-H331-H335-H336-H350-H373
Hazard Statement GHS H Statement
H225-H350-H302-H315-H319-H335
Highly flammable liquid and vapor.
May cause cancer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P311-P330-P332+P313-P363-P370+P378q-P5
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYLENE DICHLORIDE
EINECSNumber 203-458-1
RTECSNumber KI0525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only