missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloroethane, puriss. p.a., ≥99.5% (GC), Solstice

Catalog No. 60044519
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60044519 1 L Glass Bottle
60044520 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60044519 Supplier Solstice Supplier No. 035301L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Puriss. p.a.,≥99.5% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Chemical Identifiers

CAS 107-06-2
Molecular Formula C2H4Cl2
Molecular Weight (g/mol) 98.95
MDL Number MFCD00000963
InChI Key WSLDOOZREJYCGB-UHFFFAOYSA-N
Synonym ethylene dichloride, ethylene chloride, ethane, 1,2-dichloro, dutch liquid, dutch oil, ethane dichloride, glycol dichloride, aethylenchlorid, dichloro-1,2-ethane, dichloremulsion
PubChem CID 11
ChEBI CHEBI:27789
IUPAC Name 1,2-dichloroethane
SMILES ClCCCl

Specifications

Melting Point ∼ -35°C
Color Colorless
Density 1.256g/mL at 25°C
Quantity 1 L
Explosion Limit 0.156
Flash Point 13°C (55.4°F)
Ignition Point 775°F
Odor Characteristic
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 87mmHg (25°C)
Linear Formula ClCH2CH2Cl
UN Number UN1184
Beilstein 605264
Boiling Point ∼83°C
Formula Weight 98.96g/mol
Percent Purity ≥99.5% (GC)
Grade puriss. p.a.
Chemical Name or Material 1,2-Dichloroethane
Show More Show Less
Hazard Statement P201-P210-P261-P305 + P351 + P338-P311
Precautionary Statement H225-H302-H315-H319-H331-H335-H350
DOTInformation Class 3 - PG 2 - Ethylene dichloride
EINECSNumber 203-458-1
RTECSNumber KI0525000