missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloroethane, Purified, Spectrum™ Chemical

Catalog No. p-7165471
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608040 1 L Steel Can
18608042 4 L Steel Can
18608039 20 L Steel Can
18608041 200 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18608040 Supplier Spectrum Chemical Mfg Cor Supplier No. D11201LT
Only null left

Chemical Identifiers

CAS 107-06-2
Molecular Weight (g/mol) 98.96

Specifications

CAS Max % 1
Quantity 1 L
Packaging Steel Can
Boiling Point 83.5°C
Grade Purified
Chemical Name or Material 1,2-Dichloroethane

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.