missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dichloroéthane, 99+%, pur
Description
Ce produit de marque Thermo Scientific Chemicals faisait à l’origine partie du portefeuille Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit/article (SKU) original d’Acros Organics n’a pas changé dans le cadre de la transition vers la marque Thermo Scientific Chemicals.
Spécifications
Spécifications
| Nom ou substance chimique | 1,2-Dichloroethane |
| Point de fusion | -35,0 °C |
| Couleur | Incolore à jaune |
| Densité | 1.2500g/mL |
| Quantité | 2.5 L |
| Point d’éclair | 13 °C |
| Spectre infrarouge | Authentique |
| Emballage | Bouteille en verre |
| Forme physique | Liquide |
| Indice de réfraction | 1,4440 à 1,4460 |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement: May cause cancer. Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation. Highly flammable liquid and vapor. Harmful if swallowed. |
| Precautionary Statement | GHS P Statement: IF exposed or concerned: Get medical advice/attention. Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Usage réservé à la recherche