missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dichloroéthane (ACS certifié), Fisher Chemical™
Spécifications
Spécifications
| Nom ou substance chimique | 1, 2-Dichloroethane |
| Point de fusion | -35 °C |
| Couleur | Incolore |
| Densité | 1.250g/cm³ |
| Quantité | 4 L |
| Point d’éclair | 13 °C (55,4 °F) |
| Désignation | Test Réussi |
| Emballage | Bouteille en verre |
| Forme physique | Liquide |
| Résidus après évaporation | 0,002% max. |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Flammable liquid and vapor. Harmful if swallowed. Irritating to eyes, respiratory system and skin. May cause central nervous system effects. May cause adverse liver effects. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air. NFPA Health:2 Flammability:3 Instability:0 |
| DOTInformation | DOT Class 3, : Flammable Liquid |