missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloroethane, ACS, 99%, Spectrum™ Chemical

Catalog No. p-7165451
Change view
Click to view available options
Quantity:
100 mL
120 mL
500 mL
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608047 100 mL Amber Glass Bottle
18608048 120 mL Amber Glass Bottle
18608049 500 mL Amber Glass Bottle
18608043 1 L Amber Glass Bottle
18608044 4 L Amber Glass Bottle
18608045 20 L Amber Glass Bottle
18608046 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18608047 Supplier Spectrum Chemical Mfg Cor Supplier No. D1021100ML
Only null left

Chemical Identifiers

CAS 107-06-2
Molecular Weight (g/mol) 98.96

Specifications

CAS Max % 1
Quantity 100 mL
Packaging Amber Glass Bottle
Residue after Evaporation 2e-05
Titratable Acid 0.0003 meq/g
Water 0.0003
Boiling Point 83.5°C
Percent Purity 0.99
Grade Reagent
Chemical Name or Material 1,2-Dichloroethane

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.