missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dichloroethane, ACS, 99%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 200 L |
| Packaging | Amber Glass Bottle |
| Residue after Evaporation | 2e-05 |
| Titratable Acid | 0.0003 meq/g |
| Water | 0.0003 |
| Boiling Point | 83.5°C |
| Percent Purity | 0.99 |
| Grade | Reagent |
| Chemical Name or Material | 1,2-Dichloroethane |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.