missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichlorobutane 96.0+%, TCI America™

Catalog No. D034725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D034725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D034725G Supplier TCI America Supplier No. D034725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 616-21-7
Color Colorless
Boiling Point 124°C
Molecular Formula C4H8Cl2
Quantity 25 g
MDL Number MFCD00013676
UN Number 1993
Synonym butane, 1,2-dichloro, butane,2-dichloro, 1,2-butylene chloride, acmc-1b1ry, dsstox_cid_7244, dsstox_rid_78366, dsstox_gsid_27244, 1,2-dichlorobutane, pqbotznyfqwrhu-uhfffaoysa
InChI Key PQBOTZNYFQWRHU-UHFFFAOYNA-N
SMILES CCC(Cl)CCl
IUPAC Name 1,2-dichlorobutane
Molecular Weight (g/mol) 127.01
PubChem CID 12017
Formula Weight 127.01
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Dichlorobutane
Show More Show Less
EINECSNumber (2)-0061
TSCA Yes