missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichlorobutane, 97%

Catalog No. AAB2130703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2130703 1 g
AAB2130706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2130703 Supplier Thermo Scientific Chemicals Supplier No. B2130703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 616-21-7 | C4H8Cl2 | 127.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 616-21-7
Molecular Formula C4H8Cl2
Molecular Weight (g/mol) 127.01
MDL Number MFCD00013676
InChI Key PQBOTZNYFQWRHU-UHFFFAOYNA-N
Synonym butane, 1,2-dichloro, butane,2-dichloro, 1,2-butylene chloride, acmc-1b1ry, dsstox_cid_7244, dsstox_rid_78366, dsstox_gsid_27244, 1,2-dichlorobutane, pqbotznyfqwrhu-uhfffaoysa
PubChem CID 12017
IUPAC Name 1,2-dichlorobutane
SMILES CCC(Cl)CCl

Specifications

Density 1.112
Boiling Point 124°C
Flash Point 30°C (86°F)
Refractive Index 1.445
Quantity 1 g
UN Number UN1993
Beilstein 1731376
Formula Weight 127.01
Percent Purity ≥97%
Chemical Name or Material 1,2-Dichlorobutane
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 210-469-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only