missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichlorobenzene, Spectrophotometric Grade, 98%

Catalog No. p-4781744
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA32154K2 1 L
AA32154M1 4 x 1 L
AA32154K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA32154K2 Supplier Thermo Scientific Chemicals Supplier No. 032154K2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Maximum level of impurities: Evaporation residue 5ppm, H2O 0.02% | CAS: 95-50-1 | C6H4Cl2 | 146.998 g/mol

1,2-Dichlorobenzene is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 95-50-1
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 146.998
MDL Number MFCD00000535
InChI Key RFFLAFLAYFXFSW-UHFFFAOYSA-N
Synonym o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben
PubChem CID 7239
ChEBI CHEBI:35290
IUPAC Name 1,2-dichlorobenzene
SMILES C1=CC=C(C(=C1)Cl)Cl

Specifications

Melting Point °C to 18°C
Color Colorless
Density 1.306
Boiling Point 180°C
Flash Point 68°C (154°F)
Ignition Point 640°C (1184°F)
Odor Pungent
Assay Percent Range 98%
Refractive Index 1.551
Quantity 1 L
Beilstein 606078
Solubility Information Miscible with water and most of the organic solvents.
Formula Weight 147
Vapor Pressure 1.33 hPa (1mm Hg) at 20°C
Grade Spectrophotometric
Concentration or Composition (by Analyte or Components) 1, 2-Dichlorobenzene
Physical Form Liquid
Chemical Name or Material 1, 2-Dichlorobenzene, Filtered through 0.2μ filters.
Show More Show Less
Hazard Category H227-H302+H332-H315-H317-H319-H335
Hazard Statement This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P370+P378q-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 202-425-9
RTECSNumber CZ4500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only