Learn More
1,2-Dichlorobenzene, 99%
CAS: 95-50-1 | C6H4Cl2 | 146.998 g/mol
Supplier: Thermo Scientific Chemicals A138810E
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications1,2-Dichlorobenzene is widely used as a precursor to prepare 1,2-dichloro-4-nitrobenzene, which is an intermediate in the synthesis of agrochemicals. It acts as a solvent for fullerenes. It is also used in softening and removing carbon-based contamination on metal surfaces. It also used as food preservative, motor oil additive, lubricant and paint.
Solubility
Miscible with water and most of the organic solvents.
Notes
Keep away from sources of ignition. Incompatible with strong oxidizing agents.
Specifications
| 1,2-Dichlorobenzene | |
| -18°C | |
| 180°C | |
| Pungent | |
| 1.551 | |
| 2500 g | |
| 606078 | |
| o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben | |
| RFFLAFLAYFXFSW-UHFFFAOYSA-N | |
| 1,2-dichlorobenzene | |
| 7239 | |
| 147 |
| 95-50-1 | |
| 1.306 | |
| 68°C (154°F) | |
| C6H4Cl2 | |
| MFCD00000535 | |
| UN1591 | |
| 14,3056 | |
| Miscible with water and most of the organic solvents. | |
| C1=CC=C(C(=C1)Cl)Cl | |
| 146.998 | |
| CHEBI:35290 | |
| 99% |
Chemical Identifiers
| 95-50-1 | |
| 146.998 | |
| RFFLAFLAYFXFSW-UHFFFAOYSA-N | |
| 7239 | |
| 1,2-dichlorobenzene |
| C6H4Cl2 | |
| MFCD00000535 | |
| o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben | |
| CHEBI:35290 | |
| C1=CC=C(C(=C1)Cl)Cl |
Safety and Handling
GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P370+P378q-P501c
H227-H302+H332-H315-H317-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: O-DICHLOROBENZENE
EINECSNumber : 202-425-9
RTECSNumber : CZ4500000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only