missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Dichlorobenzene, 99+%, for spectroscopy
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for UV/VIS spectroscopy
Specifications
Specifications
| Alkalinity | 0.0005 meq/g max. |
| Acidity | 0.0003 meq/g max. |
| CAS Min % | 99.0 |
| Absorbance | (1 cm cell vs water), 0.01 at 380nm, 0.05 at 336nm, 0.3 at 300nm, 1 at 296nm |
| CAS Max % | 100.0 |
| Melting Point | -15.0°C |
| Density | 1.3000g/mL |
| Boiling Point | 179.0°C to 180.0°C |
| Flash Point | 67°C |
| Infrared Spectrum | Authentic |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes serious eye irritation. Harmful if swallowed. May cause respiratory irritation. Very toxic to aquatic life with long lasting effects. Causes skin irritation. Harmful if inhaled. May cause |
| Precautionary Statement | GHS P Statement Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water. If skin irritatio |
| EINECSNumber | 202-425-9 |
| RTECSNumber | CZ4500000 |
| TSCA | TSCA |
RUO – Research Use Only