missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichlorobenzene, 99%

Catalog No. p-7021519
Change view
Click to view available options
Quantity:
10,000 g
500 g
2500 g
Catalog No. Quantity
AAA1388136 500 g
AAA138810E 2500 g
AAA138810C 10,000 g
3 options

Catalog No. AAA1388136

Supplier: Thermo Scientific Chemicals A1388136

Only null left

CAS: 95-50-1 | C6H4Cl2 | 146.998 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
1,2-Dichlorobenzene is widely used as a precursor to prepare 1,2-dichloro-4-nitrobenzene, which is an intermediate in the synthesis of agrochemicals. It acts as a solvent for fullerenes. It is also used in softening and removing carbon-based contamination on metal surfaces. It also used as food preservative, motor oil additive, lubricant and paint.

Solubility
Miscible with water and most of the organic solvents.

Notes
Keep away from sources of ignition. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 95-50-1
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 146.998
MDL Number MFCD00000535
InChI Key RFFLAFLAYFXFSW-UHFFFAOYSA-N
Synonym o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben
PubChem CID 7239
ChEBI CHEBI:35290
IUPAC Name 1,2-dichlorobenzene
SMILES C1=CC=C(C(=C1)Cl)Cl

Specifications

Melting Point -18°C
Density 1.306
Boiling Point 180°C
Flash Point 68°C (154°F)
Odor Pungent
Refractive Index 1.551
Quantity 500 g
UN Number UN1591
Beilstein 606078
Merck Index 14,3056
Solubility Information Miscible with water and most of the organic solvents.
Formula Weight 147
Percent Purity 99%
Chemical Name or Material 1,2-Dichlorobenzene
Show More Show Less
Hazard Category H227-H302+H332-H315-H317-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: O-DICHLOROBENZENE
EINECSNumber 202-425-9
RTECSNumber CZ4500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only