missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromotetrachloroethane, 97%

Catalog No. AC158460250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158460250 25 g Glass bottle
AC158461000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC158460250 Supplier Thermo Scientific Chemicals Supplier No. 158460250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 630-25-1 | C2Br2Cl4 | 325.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 630-25-1
Molecular Formula C2Br2Cl4
Molecular Weight (g/mol) 325.63
MDL Number MFCD00000780
InChI Key WJUKOGPNGRUXMG-UHFFFAOYSA-N
Synonym 1,2-dibromotetrachloroethane, dbtce, sym-dibromotetrachloroethane, ethane, 1,2-dibromo-1,1,2,2-tetrachloro, unii-0sgg217efr, 0sgg217efr, 1,2-dibrom-1,1,2,2-tetrachlorethan, brccl2ccl2br, 1,2-dibromotetrachloroethan, 1,2-dibrom-tetrachlorethane
PubChem CID 69426
IUPAC Name 1,2-dibromo-1,1,2,2-tetrachloroethane
SMILES C(C(Cl)(Cl)Br)(Cl)(Cl)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 220.0°C to 222.0°C
Color White to Beige
Density 2.7100g/mL
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrCCl2CCl2Br
Quantity 25 g
Beilstein 01, 93
Fieser 01,209
Specific Gravity 2.71
Solubility Information Solubility in water: insoluble
Formula Weight 325.63
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 1, 2-Dibromotetrachloroethane, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-136-7
RTECSNumber KH9366000
TSCA TSCA

RUO – Research Use Only