missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromotetrachloroethane, 97%

Catalog No. p-7024991
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2027614 25 g
AAB2027622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2027614 Supplier Thermo Scientific Chemicals Supplier No. B2027614
Only null left

CAS: 630-25-1 | C2Br2Cl4 | 325.63 g/mol

1,2-dibromotetrachloroethane is used as a fire-proofing agent. It is actively involved in the conversion of sulfones to alkenes by Ramberg-Backlund rearrangement. It is an effective replacement for ozone depleting solvents like dibromodifluoromethane and tetrabromodifluoroethane. It is also utilized as a brominating reagent. In analytical chemistry, it is used in quantitative determination of perhalogenated compounds by high-performance liquid chromatography (HPLC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-dibromotetrachloroethane is used as a fire-proofing agent. It is actively involved in the conversion of sulfones to alkenes by Ramberg-Backlund rearrangement. It is an effective replacement for ozone depleting solvents like dibromodifluoromethane and tetrabromodifluoroethane. It is also utilized as a brominating reagent. In analytical chemistry, it is used in quantitative determination of perhalogenated compounds by high-performance liquid chromatography (HPLC).

Solubility
Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 630-25-1
Molecular Formula C2Br2Cl4
Molecular Weight (g/mol) 325.63
MDL Number MFCD00000780
InChI Key WJUKOGPNGRUXMG-UHFFFAOYSA-N
Synonym 1,2-dibromotetrachloroethane, dbtce, sym-dibromotetrachloroethane, ethane, 1,2-dibromo-1,1,2,2-tetrachloro, unii-0sgg217efr, 0sgg217efr, 1,2-dibrom-1,1,2,2-tetrachlorethan, brccl2ccl2br, 1,2-dibromotetrachloroethan, 1,2-dibrom-tetrachlorethane
PubChem CID 69426
IUPAC Name 1,2-dibromo-1,1,2,2-tetrachloroethane
SMILES C(C(Cl)(Cl)Br)(Cl)(Cl)Br

Specifications

Melting Point ∼220°C (decomposition)
Density 2.71
Quantity 25 g
Beilstein 1699471
Solubility Information Insoluble in water.
Formula Weight 325.65
Percent Purity 98%
Chemical Name or Material 1,2-Dibromotetrachloroethane
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 211-136-7
RTECSNumber KH9366000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only