missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromohexafluoropropane, 95%

Catalog No. AAA1710314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1710314 25 g
AAA1710322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1710314 Supplier Thermo Scientific Chemicals Supplier No. A1710314

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 661-95-0 | C3Br2F6 | 309.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 661-95-0
Molecular Formula C3Br2F6
Molecular Weight (g/mol) 309.83
MDL Number MFCD00013568
InChI Key KTULQNFKNLFOHL-UHFFFAOYNA-N
Synonym 1,2-dibromohexafluoropropane, propane, 1,2-dibromo-1,1,2,3,3,3-hexafluoro
PubChem CID 69579
IUPAC Name 1,2-dibromo-1,1,2,3,3,3-hexafluoropropane
SMILES FC(F)(F)C(F)(Br)C(F)(F)Br

Specifications

Density 2.169
Boiling Point 71°C to 72°C
Quantity 25 g
Formula Weight 309.84
Percent Purity 95%
Chemical Name or Material 1,2-Dibromohexafluoropropane
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 211-550-8
RTECSNumber TX8800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only