missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromo-2-methylpropane, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 406490250

 View more versions of this product

Catalog No. AC406490250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

1, 2-Dibromo-2-methylpropane
594-34-3
100.0
150.0°C
Authentic
Glass bottle
1.5083 to 1.5103
MFCD00000126
1.79
Solubility in water: slightly soluble
CC(C)(CBr)Br
215.92
215.92
98%
97.5
1.7900g/mL
>110°C
97.5% min. (GC)
C4H8Br2
(CH3)2CBrCH2Br
25g
isobutylene bromide, 1,2-dibromoisobutane, acmc-1aolc, propane,2-dibromo-2-methyl, propane,1,2-dibromo-2-methyl, 1,2-dibromo-2-methylpropane
SDTXSEXYPROZSZ-UHFFFAOYSA-N
1,2-dibromo-2-methylpropane
222657
98%

Chemical Identifiers

594-34-3
215.92
SDTXSEXYPROZSZ-UHFFFAOYSA-N
222657
CC(C)(CBr)Br
C4H8Br2
MFCD00000126
isobutylene bromide, 1,2-dibromoisobutane, acmc-1aolc, propane,2-dibromo-2-methyl, propane,1,2-dibromo-2-methyl, 1,2-dibromo-2-methylpropane
1,2-dibromo-2-methylpropane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Suspected of causing cancer.

GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-835-7

RTECSNumber : TX8810000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only