missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibenzoylethane, 98+%

Catalog No. AAL0865206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0865206 5 g
AAL0865203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0865206 Supplier Thermo Scientific Chemicals Supplier No. L0865206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 495-71-6 | C16H14O2 | 238.286 g/mol

1,2-Dibenzoylethane is effective in inhibiting the mammary DMBA-DNA adduct formation. This inhibitory effect on mammary DNA adduct formation was associated with increased liver activities of glutathione S-transferase, QR, and 7-ethoxyresorufin-O-deethylase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Dibenzoylethane is effective in inhibiting the mammary DMBA-DNA adduct formation. This inhibitory effect on mammary DNA adduct formation was associated with increased liver activities of glutathione S-transferase, QR, and 7-ethoxyresorufin-O-deethylase.

Solubility
Soluble in acetone. Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 495-71-6
Molecular Formula C16H14O2
Molecular Weight (g/mol) 238.286
MDL Number MFCD00037818
InChI Key OSWWFLDIIGGSJV-UHFFFAOYSA-N
Synonym 1,2-dibenzoylethane, 1,4-diphenyl-1,4-butanedione, 1,4-butanedione, 1,4-diphenyl, biphenacyl, diphenacyl, 2,2-biacetophenone, acmc-1bd9f, ethane, 1,2-dibenzoyl, succinophenone diphenacyl, cambridge id 5102527
PubChem CID 136322
IUPAC Name 1,4-diphenylbutane-1,4-dione
SMILES C1=CC=C(C=C1)C(=O)CCC(=O)C2=CC=CC=C2

Specifications

Melting Point 144°C to 148°C
Boiling Point 260°C (15 mmHg)
Quantity 5 g
Beilstein 782937
Solubility Information Soluble in acetone. Insoluble in water.
Formula Weight 238.29
Percent Purity ≥98%
Chemical Name or Material 1,2-Dibenzoylethane
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only