missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diaminopropane 98.0+%, TCI America™

Catalog No. p-7126042
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D011325ML 25 mL
D0113500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D011325ML Supplier TCI America Supplier No. D011325ML

Chemical Identifiers

CAS 78-90-0
Molecular Formula C3H10N2
Molecular Weight (g/mol) 74.13
MDL Number MFCD00008089
InChI Key AOHJOMMDDJHIJH-UHFFFAOYNA-N
Synonym 1,2-diaminopropane, propylenediamine, 1,2-propanediamine, propylene diamine, 2,3-diaminopropane, 1,2-propylenediamine, 1,2 diaminopropane, ccris 4863, 1-methylethylenediamine, 2-aminopropylamine
PubChem CID 6567
ChEBI CHEBI:30630
IUPAC Name propane-1,2-diamine
SMILES CC(N)CN

Specifications

Color Yellow
Boiling Point 120°C
Quantity 25 mL
UN Number 2258
Formula Weight 74.13
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1,2-Diaminopropane
EINECSNumber (2)-0149
RTECSNumber TX6650000
TSCA Yes