missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diaminopropane, 99%

Catalog No. p-7027604
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB24489AE 100 mL
AAB24489AP 500 mL
AAB24489AU 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB24489AE Supplier Thermo Scientific Chemicals Supplier No. B24489AE
Only null left

CAS: 78-90-0 | C3H10N2 | 74.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78-90-0
Molecular Formula C3H10N2
Molecular Weight (g/mol) 74.13
MDL Number MFCD00008089
InChI Key AOHJOMMDDJHIJH-UHFFFAOYNA-N
Synonym 1,2-diaminopropane, propylenediamine, 1,2-propanediamine, propylene diamine, 2,3-diaminopropane, 1,2-propylenediamine, 1,2 diaminopropane, ccris 4863, 1-methylethylenediamine, 2-aminopropylamine
PubChem CID 6567
ChEBI CHEBI:30630
IUPAC Name propane-1,2-diamine
SMILES CC(N)CN

Specifications

Melting Point ∼-37°C
Density 0.869
Boiling Point 119°C to 120°C
Flash Point 34°C (93°F)
Odor Characteristic
Refractive Index 1.446
Quantity 100 mL
UN Number UN2258
Beilstein 605274
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,7852
Formula Weight 74.13
Percent Purity 99%
Chemical Name or Material 1,2-Diaminopropane
Show More Show Less
Hazard Category H226-H302-H311-H314-H317-H334
Hazard Statement GHS H Statement
H314-H226-H302-H312
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P272-P280-P285-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P374-P380-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: 1,2-PROPYLENEDIAMINE
EINECSNumber 201-155-9
RTECSNumber TX6650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only