missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diaminocyclohexanetetraacetic acid monohydrate, Honeywell Fluka™

for complexometry,≥98.5%

$292.15 - $954.35

Chemical Identifiers

CAS 145819-99-4
Molecular Formula C14H24N2O9
Molecular Weight (g/mol) 364.351
MDL Number MFCD00150952
InChI Key VASZYFIKPKYGNC-UHFFFAOYSA-N
Synonym cdta hydrate, glycine, n,n'-1,2-cyclohexanediylbis n-carboxymethyl-, monohydrate, trans-1,2-cyclohexanediamine-n,n,n',n'-tetraacetic acid monohydrate, acmc-20n4nc, 1,2-diaminocyclohexanetetraacetic acid monohydrate, trans-1,2-cyclohexanediamine-n,n,n',n-tetraacetic acid, 2,2',2,2'-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid hydrate, 2,2',2,2'-cyclohexane-1,2-diyldinitrilo tetraacetic acid-water 1/1, 2-2-bis carboxymethyl amino cyclohexyl-carboxymethyl amino acetic acid hydrate, 1,2-diaminocyclohexanetetraacetic acid monohydrate for complexometry, inverted exclamation marky
PubChem CID 18674933
IUPAC Name 2-[[2-[bis(carboxymethyl)amino]cyclohexyl]-(carboxymethyl)amino]acetic acid;hydrate
SMILES C1CCC(C(C1)N(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6002543
View Documents
Honeywell-Fluka
3458825G
25 g
Each for $292.15
Only null left
 
6002542
View Documents
Honeywell-Fluka
34588100G
100 g
Each for $954.35
Only null left
 
Description

Description

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

Specifications

Chemical Identifiers

145819-99-4
364.351
VASZYFIKPKYGNC-UHFFFAOYSA-N
18674933
C1CCC(C(C1)N(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.O
C14H24N2O9
MFCD00150952
cdta hydrate, glycine, n,n'-1,2-cyclohexanediylbis n-carboxymethyl-, monohydrate, trans-1,2-cyclohexanediamine-n,n,n',n'-tetraacetic acid monohydrate, acmc-20n4nc, 1,2-diaminocyclohexanetetraacetic acid monohydrate, trans-1,2-cyclohexanediamine-n,n,n',n-tetraacetic acid, 2,2',2,2'-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid hydrate, 2,2',2,2'-cyclohexane-1,2-diyldinitrilo tetraacetic acid-water 1/1, 2-2-bis carboxymethyl amino cyclohexyl-carboxymethyl amino acetic acid hydrate, 1,2-diaminocyclohexanetetraacetic acid monohydrate for complexometry, inverted exclamation marky
2-[[2-[bis(carboxymethyl)amino]cyclohexyl]-(carboxymethyl)amino]acetic acid;hydrate

Specifications

145819-99-4
MFCD00150952
NONH for all modes of transport
cdta hydrate, glycine, n,n'-1,2-cyclohexanediylbis n-carboxymethyl-, monohydrate, trans-1,2-cyclohexanediamine-n,n,n',n'-tetraacetic acid monohydrate, acmc-20n4nc, 1,2-diaminocyclohexanetetraacetic acid monohydrate, trans-1,2-cyclohexanediamine-n,n,n',n-tetraacetic acid, 2,2',2,2'-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid hydrate, 2,2',2,2'-cyclohexane-1,2-diyldinitrilo tetraacetic acid-water 1/1, 2-2-bis carboxymethyl amino cyclohexyl-carboxymethyl amino acetic acid hydrate, 1,2-diaminocyclohexanetetraacetic acid monohydrate for complexometry, inverted exclamation marky
C1CCC(C(C1)N(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.O
364.351
364.35g/mol
1,2-Diaminocyclohexanetetraacetic acid monohydrate
C14H24N2O9
25 g
6884761
VASZYFIKPKYGNC-UHFFFAOYSA-N
2-[[2-[bis(carboxymethyl)amino]cyclohexyl]-(carboxymethyl)amino]acetic acid;hydrate
18674933
≥98.5%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'