missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diaminocyclohexanetetraacetic acid disodium salt solution, Solstice

Catalog No. p-7072155
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002597 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6002597 Supplier Solstice Supplier No. 351031L

Volumetric, Reag. Ph. Eur., 0.1 M DCTA-Na4, for complexometry

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

Chemical Identifiers

CAS 5786-78-7
Molecular Formula C14H20N2Na2O8
Molecular Weight (g/mol) 390.3
InChI Key TVTLKUMLAQUFAS-UHFFFAOYSA-L
Synonym sodium 1,2-diaminocyclohexane tetraacetate, 1,2-diaminocyclohexenetetra-acetate of sodium, acetic acid, 1,2-cyclohexylenedinitrilo tetra-, sodium salt, glycine, n,n'-1,2-cyclohexanediylbis n-carboxymethyl-, disodium salt, sodium n'-1,2-cyclohexanediylbis n-carboxymethyl glycinate, glycine, n,n'-1,2-cyclohexanediylbis n-carboxymethyl-, sodium salt 1:2, cdta disodium salt, disodium 2-carboxymethyl-2-carboxymethyl-2-oxido-2-oxoethyl amino cyclohexyl amino acetate
PubChem CID 24230
IUPAC Name disodium;2-[[2-[carboxylatomethyl(carboxymethyl)amino]cyclohexyl]-(carboxymethyl)amino]acetate
SMILES C1CCC(C(C1)N(CC(=O)O)CC(=O)[O-])N(CC(=O)O)CC(=O)[O-].[Na+].[Na+]

Specifications

Quantity 1 L
UN Number NONH for all modes of transport
Beilstein 6884761
Formula Weight 390.3g/mol
Chemical Name or Material 1,2-Diaminocyclohexanetetraacetic acid disodium salt solution