missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Diacetoxypropane 98.0+%, TCI America™
$29.83 - $67.11
Chemical Identifiers
| CAS | 623-84-7 |
|---|---|
| Molecular Formula | C7H12O4 |
| Molecular Weight (g/mol) | 160.17 |
| MDL Number | MFCD00026201 |
| InChI Key | MLHOXUWWKVQEJB-UHFFFAOYNA-N |
| Synonym | propylene glycol diacetate, 1,2-diacetoxypropane, propylene diacetate, propylene acetate, methylethylene acetate, 1,2-propylene glycol diacetate, 1,2-propanediol, diacetate, methylethylene diacetate, propyleneglycol diacetate, 1,2-propanediol diacetate |
| PubChem CID | 12198 |
| IUPAC Name | 1-(acetyloxy)propan-2-yl acetate |
| SMILES | CC(COC(C)=O)OC(C)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P090225ML
|
TCI America
P090225ML |
25 mL |
Each for $29.83
|
|
|||||
|
P0902500ML
|
TCI America
P0902500ML |
500 mL |
Each for $67.11
|
|
|||||
Chemical Identifiers
| 623-84-7 | |
| 160.17 | |
| MLHOXUWWKVQEJB-UHFFFAOYNA-N | |
| 12198 | |
| CC(COC(C)=O)OC(C)=O |
| C7H12O4 | |
| MFCD00026201 | |
| propylene glycol diacetate, 1,2-diacetoxypropane, propylene diacetate, propylene acetate, methylethylene acetate, 1,2-propylene glycol diacetate, 1,2-propanediol, diacetate, methylethylene diacetate, propyleneglycol diacetate, 1,2-propanediol diacetate | |
| 1-(acetyloxy)propan-2-yl acetate |
Specifications
| 623-84-7 | |
| Colorless | |
| C7H12O4 | |
| 25 mL | |
| MLHOXUWWKVQEJB-UHFFFAOYNA-N | |
| 1-(acetyloxy)propan-2-yl acetate | |
| 12198 | |
| ≥98.0% (GC) | |
| 1,2-Diacetoxypropane |
| -31°C | |
| 186°C | |
| MFCD00026201 | |
| propylene glycol diacetate, 1,2-diacetoxypropane, propylene diacetate, propylene acetate, methylethylene acetate, 1,2-propylene glycol diacetate, 1,2-propanediol, diacetate, methylethylene diacetate, propyleneglycol diacetate, 1,2-propanediol diacetate | |
| CC(COC(C)=O)OC(C)=O | |
| 160.17 | |
| 160.17 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0665
RTECSNumber : TY4900000
TSCA : Yes