missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diacetoxypropane 98.0+%, TCI America™

Catalog No. P090225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P090225ML 25 mL
P0902500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P090225ML Supplier TCI America Supplier No. P090225ML
Only null left

Chemical Identifiers

CAS 623-84-7
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.17
MDL Number MFCD00026201
InChI Key MLHOXUWWKVQEJB-UHFFFAOYNA-N
Synonym propylene glycol diacetate, 1,2-diacetoxypropane, propylene diacetate, propylene acetate, methylethylene acetate, 1,2-propylene glycol diacetate, 1,2-propanediol, diacetate, methylethylene diacetate, propyleneglycol diacetate, 1,2-propanediol diacetate
PubChem CID 12198
IUPAC Name 1-(acetyloxy)propan-2-yl acetate
SMILES CC(COC(C)=O)OC(C)=O

Specifications

Melting Point -31°C
Color Colorless
Boiling Point 186°C
Quantity 25 mL
Formula Weight 160.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Diacetoxypropane
EINECSNumber (2)-0665
RTECSNumber TY4900000
TSCA Yes