missing translation for 'onlineSavingsMsg'
Learn More

1,2-Di(p-tolyl)ethane, 98%

Catalog No. AAL0887603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0887603 1 g
AAL0887606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0887603 Supplier Thermo Scientific Chemicals Supplier No. L0887603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 538-39-6 | C16H18 | 210.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 538-39-6
Molecular Formula C16H18
Molecular Weight (g/mol) 210.32
MDL Number MFCD00026025
InChI Key XCCQFUHBIRHLQT-UHFFFAOYSA-N
Synonym 1,2-di p-tolyl ethane, 4,4'-dimethylbibenzyl, 1,2-di-p-tolylethane, unii-ua2zyh96ih, bibenzyl, 4,4'-dimethyl, 1-methyl-4-2-4-methylphenyl ethyl benzene, ua2zyh96ih, 1,2-bis 4-methylphenyl ethane, benzene, 1,1'-1,2-ethanediyl bis 4-methyl
PubChem CID 10854
IUPAC Name 1-methyl-4-[2-(4-methylphenyl)ethyl]benzene
SMILES CC1=CC=C(C=C1)CCC2=CC=C(C=C2)C

Specifications

Melting Point 80°C to 82°C
Boiling Point 296°C to 298°C (730 mmHg)
Quantity 1 g
Beilstein 1909630
Merck Index 14,3380
Formula Weight 210.32
Percent Purity 98%
Chemical Name or Material 1,2-Di(p-tolyl)ethane
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
RTECSNumber YT4380000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only