missing translation for 'onlineSavingsMsg'
Learn More

1,2-Di(4-pyridyl)ethylene 98.0+%, TCI America™

Catalog No. p-7126118
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D027610G 10 g
D027625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D027610G Supplier TCI America Supplier No. D027610G

Chemical Identifiers

CAS 13362-78-2
Molecular Formula C12H10N2
Molecular Weight (g/mol) 182.226
MDL Number MFCD00006448
InChI Key MGFJDEHFNMWYBD-OWOJBTEDSA-N
Synonym 4,4'-vinylenedipyridine, 1,2-di 4-pyridyl ethylene, 1,2-bis 4-pyridyl ethylene, 1,2-di pyridin-4-yl ethene, trans-1,2-bis 4-pyridyl ethylene, 1,2-di-4-pyridylethene, e-1,2-di pyridin-4-yl ethene, pyridine, 4,4'-1e-1,2-ethenediylbis, 4-4-pyridylvinyl pyridine, pyridine, 4,4'-1,2-ethenediyl bis-, e
PubChem CID 776222
IUPAC Name 4-[(E)-2-pyridin-4-ylethenyl]pyridine
SMILES C1=CN=CC=C1C=CC2=CC=NC=C2

Specifications

Melting Point 151°C
Quantity 10 g
Formula Weight 182.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Di(4-pyridyl)ethylene