missing translation for 'onlineSavingsMsg'
Learn More

1,2-Di(4-pyridyl)ethane 98.0+%, TCI America™

Catalog No. p-7127893
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37521G 1 g
D37525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37521G Supplier TCI America Supplier No. D37521G
Only null left

Chemical Identifiers

CAS 4916-57-8
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.242
MDL Number MFCD00006451
InChI Key DQRKTVIJNCVZAX-UHFFFAOYSA-N
Synonym 1,2-bis 4-pyridyl ethane, 1,2-di pyridin-4-yl ethane, 4,4'-ethylenedipyridine, 1,2-di 4-pyridyl ethane, 4,4'-ethane-1,2-diyldipyridine, unii-wo1evc302b, pyridine, 4,4'-1,2-ethanediyl bis, wo1evc302b, chembl70083, 4-2-pyridin-4-ylethyl pyridine
PubChem CID 78630
IUPAC Name 4-(2-pyridin-4-ylethyl)pyridine
SMILES C1=CN=CC=C1CCC2=CC=NC=C2

Specifications

Melting Point 112°C
Color White-Yellow
Boiling Point 174°C
Quantity 1 g
Formula Weight 184.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Di(4-pyridyl)ethane