missing translation for 'onlineSavingsMsg'
Learn More

1,2-Cyclohexanedione dioxime, 97%

Catalog No. p-7023345
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1678806 5 g
AAA1678814 25 g
AAA1678822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1678806 Supplier Thermo Scientific Chemicals Supplier No. A1678806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 492-99-9 | C6H10N2O2 | 142.16 g/mol

1,2-Cyclohexanedione dioxime acts as a chelating ligand forming complexes with many transition metal ions like Co, Fe and Ni. It is used in photometric detection of metal ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Cyclohexanedione dioxime acts as a chelating ligand forming complexes with many transition metal ions like Co, Fe and Ni. It is used in photometric detection of metal ions.

Solubility
Soluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 492-99-9
Molecular Formula C6H10N2O2
Molecular Weight (g/mol) 142.16
MDL Number MFCD00001663
InChI Key CUNNCKOPAWXYDX-KQQUZDAGSA-N
Synonym 1,2-cyclohexanedione dioxime, nioxime, 1z,2z-cyclohexane-1,2-dione dioxime, nioxim, 1,2-cyclohexanedione, dioxime, 1,2-bis hydroxyimino cyclohexane, unii-g1qt7su5a8, cyclohexane-1,2-dione dioxime, 1,2-cyclohexanedione, 1,2-dioxime, g1qt7su5a8
PubChem CID 10300
IUPAC Name N-(2-nitrosocyclohexen-1-yl)hydroxylamine
SMILES O\N=C1/CCCC/C/1=N\O

Specifications

Melting Point 181°C to 188°C (decomposition)
Quantity 5 g
Beilstein 1909047
Merck Index 14,6560
Solubility Information Soluble in water.
Formula Weight 142.16
Percent Purity 97%
Chemical Name or Material 1,2-Cyclohexanedione dioxime
EINECSNumber 207-769-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only