missing translation for 'onlineSavingsMsg'
Learn More

1,2-Cyclohexanedione, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 111162500

 View more versions of this product

Catalog No. AC111162500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

1, 2-Cyclohexanedione
193.0°C to 195.0°C
Authentic
Glass Bottle
1.5180 to 1.5200
250g
02,93
Solubility in water: soluble. Other solubilities: soluble in ethanol and diethyl ether
C1CCC(=O)C(=O)C1
112.13
CHEBI:41674
98%
765-87-7
84°C
97.5% min. (GC)
C6H8O2
C6H8(=O)2
07,II,526
1,2-cyclohexanedione, 1,2-dioxocyclohexane, cyclohexanedione, 1,2-cyclohexadione, cyclohexan-1,2-dione, ccris 6296, unii-75c1ovw0fj, 75c1ovw0fj, 1,2-cyclohexanedione,ketone form, dihydrocatechol
OILAIQUEIWYQPH-UHFFFAOYSA-N
cyclohexane-1,2-dione
13006
112.13

Chemical Identifiers

765-87-7
112.13
1,2-cyclohexanedione, 1,2-dioxocyclohexane, cyclohexanedione, 1,2-cyclohexadione, cyclohexan-1,2-dione, ccris 6296, unii-75c1ovw0fj, 75c1ovw0fj, 1,2-cyclohexanedione,ketone form, dihydrocatechol
CHEBI:41674
C1CCC(=O)C(=O)C1
C6H8O2
OILAIQUEIWYQPH-UHFFFAOYSA-N
13006
cyclohexane-1,2-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 212-155-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only