missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Cyclohexanedicarboxylic anhydride, cis + trans, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 32°C to 34°C |
| Boiling Point | 158°C (17 mmHg) |
| Flash Point | 275°C (527°F) |
| Quantity | 100 g |
| Beilstein | 83213 |
| Sensitivity | Moisture sensitive |
| Formula Weight | 154.17 |
| Percent Purity | 97% |
| Chemical Name or Material | 1,2-Cyclohexanedicarboxylic anhydride, Cis + Trans |
Safety and Handling
| Hazard Category | H317-H318-H334 |
| Hazard Statement | GHS H Statement H334-H318-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes serious eye damage. May cause an allergic skin reaction. |
| Precautionary Statement | P261-P272-P280-P285-P302+P352-P304+P341-P305+P351+P338-P310-P333+P313-P342+P311-P363-P501c |
| EINECSNumber | 201-604-9 |
| RTECSNumber | NP6895168 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only