missing translation for 'onlineSavingsMsg'
Learn More

1,2-Cyclohexanedicarboxylic anhydride, cis + trans, 97%

Catalog No. AAL2000122
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2000122 100 g
AAL2000136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2000122 Supplier Thermo Scientific Chemicals Supplier No. L2000122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 85-42-7 | C8H10O3 | 154.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 85-42-7
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.17
MDL Number MFCD00064863,MFCD00674195
InChI Key MUTGBJKUEZFXGO-UHFFFAOYNA-N
Synonym hexahydrophthalic anhydride, hexahydroisobenzofuran-1,3-dione, hhpa, 1,2-cyclohexanedicarboxylic anhydride, lekutherm hardener h, hexahydrophthalic acid anhydride, 1,3-isobenzofurandione, hexahydro, araldite ht 907, cyclohexane-1,2-dicarboxylic anhydride, 1,2-cyclohexanedicarboxylic acid anhydride
PubChem CID 85689
ChEBI CHEBI:103210
IUPAC Name 3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione
SMILES O=C1OC(=O)C2CCCCC12

Specifications

Melting Point 32°C to 34°C
Boiling Point 158°C (17 mmHg)
Flash Point 275°C (527°F)
Quantity 100 g
Beilstein 83213
Sensitivity Moisture sensitive
Formula Weight 154.17
Percent Purity 97%
Chemical Name or Material 1,2-Cyclohexanedicarboxylic anhydride, Cis + Trans
Hazard Category H317-H318-H334
Hazard Statement GHS H Statement
H334-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280-P285-P302+P352-P304+P341-P305+P351+P338-P310-P333+P313-P342+P311-P363-P501c
EINECSNumber 201-604-9
RTECSNumber NP6895168
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only