missing translation for 'onlineSavingsMsg'
Learn More

1,2-Cyclohexanediamine (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. p-7126119
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D027725ML 25 mL
D0277500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D027725ML Supplier TCI America Supplier No. D027725ML
Only null left

Chemical Identifiers

CAS 694-83-7
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00001491
InChI Key SSJXIUAHEKJCMH-UHFFFAOYSA-N
Synonym 1,2-cyclohexanediamine, 1,2-diaminocyclohexane, 1,2-cylohexanediamine, cyclohex-1,2-ylenediamine, 1r,2r---1,2-diamino cyclohexane, dsstox_cid_7301, dsstox_rid_78396, dsstox_gsid_27301, +/--trans-1,2-diaminocyclohexane, cis-1,2-cyclohexandiamine
PubChem CID 4610
IUPAC Name cyclohexane-1,2-diamine
SMILES C1CCC(C(C1)N)N

Specifications

Color Yellow
Boiling Point 376°C
Quantity 25 mL
UN Number 2735
Formula Weight 114.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Cyclohexanediamine (cis- and trans- mixture)
EINECSNumber (3)-4230
RTECSNumber GU8749500
TSCA Yes