missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Cyanoethyl)-2-ethyl-4-methylimidazole (contains 5-methyl isomer) 70.0+%, TCI America™

Catalog No. C148625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C148625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C148625G Supplier TCI America Supplier No. C148625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23996-25-0
Molecular Formula C9H13N3
Molecular Weight (g/mol) 163.224
InChI Key UIDDPPKZYZTEGS-UHFFFAOYSA-N
Synonym 3-(2-Ethyl-4-methyl-1-imidazolyl)propionitrile
PubChem CID 90327
IUPAC Name 3-(2-ethyl-4-methylimidazol-1-yl)propanenitrile
SMILES CCC1=NC(=CN1CCC#N)C

Specifications

Color Yellow
pH 10.36 (1% H2O soln.)
Quantity 25 g
UN Number 3276
Formula Weight 163.22
Percent Purity ≥70.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(2-Cyanoethyl)-2-ethyl-4-methylimidazole (contains 5-methyl isomer)
EINECSNumber (5)-0401
TSCA Yes