missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Chlorophenyl)piperazine 98.0+%, TCI America™
$137.94 - $359.76
Chemical Identifiers
| CAS | 39512-50-0 |
|---|---|
| Molecular Formula | C10H13ClN2 |
| Molecular Weight (g/mol) | 196.678 |
| MDL Number | MFCD00040727 |
| InChI Key | PWZDJIUQHUGFRJ-UHFFFAOYSA-N |
| Synonym | 1-2-chlorophenyl piperazine, 1-2-chloro-phenyl-piperazine, piperazine, 1-2-chlorophenyl, chembl9841, 1-o-chlorophenyl piperazine, 2-chlorophenyl piperazine, chlorphenylpiperazin, pubchem14571, acmc-1aif1 |
| PubChem CID | 415628 |
| IUPAC Name | 1-(2-chlorophenyl)piperazine |
| SMILES | C1CN(CCN1)C2=CC=CC=C2Cl |
Chemical Identifiers
| 39512-50-0 | |
| 196.678 | |
| PWZDJIUQHUGFRJ-UHFFFAOYSA-N | |
| 415628 | |
| C1CN(CCN1)C2=CC=CC=C2Cl |
| C10H13ClN2 | |
| MFCD00040727 | |
| 1-2-chlorophenyl piperazine, 1-2-chloro-phenyl-piperazine, piperazine, 1-2-chlorophenyl, chembl9841, 1-o-chlorophenyl piperazine, 2-chlorophenyl piperazine, chlorphenylpiperazin, pubchem14571, acmc-1aif1 | |
| 1-(2-chlorophenyl)piperazine |
Specifications
| 39512-50-0 | |
| 135°C | |
| MFCD00040727 | |
| 1-2-chlorophenyl piperazine, 1-2-chloro-phenyl-piperazine, piperazine, 1-2-chlorophenyl, chembl9841, 1-o-chlorophenyl piperazine, 2-chlorophenyl piperazine, chlorphenylpiperazin, pubchem14571, acmc-1aif1 | |
| C1CN(CCN1)C2=CC=CC=C2Cl | |
| 196.678 | |
| 196.68 | |
| Liquid |
| Yellow | |
| C10H13ClN2 | |
| 5 g | |
| PWZDJIUQHUGFRJ-UHFFFAOYSA-N | |
| 1-(2-chlorophenyl)piperazine | |
| 415628 | |
| ≥98.0% (GC,T) | |
| 1-(2-Chlorophenyl)piperazine |
Safety and Handling
TSCA : No