missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Chloroethyl)pyrrolidine Hydrochloride 98.0+%, TCI America™

Catalog No. C120225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C120225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C120225G Supplier TCI America Supplier No. C120225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7250-67-1
Molecular Formula C6H13Cl2N
Molecular Weight (g/mol) 170.077
MDL Number MFCD00012718
InChI Key FSNGFFWICFYWQC-UHFFFAOYSA-N
Synonym 1-2-chloroethyl pyrrolidine hydrochloride, n-2-chloroethyl pyrrolidine hydrochloride, 2-chloroethylpyrrolidine hydrochloride, 2-pyrrolidinoethyl chloride hydrochloride, pyrrolidine, 1-2-chloroethyl-, hydrochloride, unii-yyq1h343r5, pyrrolidinoethyl chloride, hydrochloride, 1-2-chloroethyl pyrrolidine.hcl, 1-2-chloroethyl pyrrolidine hcl, 2-chloroethylpyrrolidinehydrochloride
PubChem CID 81668
IUPAC Name 1-(2-chloroethyl)pyrrolidine;hydrochloride
SMILES C1CCN(C1)CCCl.Cl

Specifications

Melting Point 172°C
Color White-Yellow
Boiling Point 237°C
Quantity 25 g
Formula Weight 170.08
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2-Chloroethyl)pyrrolidine Hydrochloride
RTECSNumber UY0703150
TSCA No