missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Chloroethyl)pyrrolidine hydrochloride, 98%

Catalog No. p-7026303
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2223614 25 g
AAB2223622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2223614 Supplier Thermo Scientific Chemicals Supplier No. B2223614

CAS: 7250-67-1 | C6H13Cl2N | 170.077 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7250-67-1
Molecular Formula C6H13Cl2N
Molecular Weight (g/mol) 170.077
MDL Number MFCD00012718
InChI Key FSNGFFWICFYWQC-UHFFFAOYSA-N
Synonym 1-2-chloroethyl pyrrolidine hydrochloride, n-2-chloroethyl pyrrolidine hydrochloride, 2-chloroethylpyrrolidine hydrochloride, 2-pyrrolidinoethyl chloride hydrochloride, pyrrolidine, 1-2-chloroethyl-, hydrochloride, unii-yyq1h343r5, pyrrolidinoethyl chloride, hydrochloride, 1-2-chloroethyl pyrrolidine.hcl, 1-2-chloroethyl pyrrolidine hcl, 2-chloroethylpyrrolidinehydrochloride
PubChem CID 81668
IUPAC Name 1-(2-chloroethyl)pyrrolidine;hydrochloride
SMILES C1CCN(C1)CCCl.Cl

Specifications

Melting Point 168°C to 173°C
Quantity 25 g
Beilstein 3677061
Sensitivity Hygroscopic
Formula Weight 170.08
Percent Purity 98%
Chemical Name or Material 1-(2-Chloroethyl)pyrrolidine hydrochloride
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 230-660-7
RTECSNumber UY0703150
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only