missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Chloroethyl)piperidine Hydrochloride 98.0+%, TCI America™

Catalog No. C1073500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1073500G 500 g
C107325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1073500G Supplier TCI America Supplier No. C1073500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2008-75-5
Molecular Formula C7H15Cl2N
Molecular Weight (g/mol) 184.104
MDL Number MFCD00012837
InChI Key VFLQQZCRHPIGJU-UHFFFAOYSA-N
Synonym 1-2-chloroethyl piperidine hydrochloride, 2-piperidinoethylchloride hydrochloride, 2-chloroethylpiperidine hydrochloride, n-chloroethyl piperidine hydrochloride, piperidine, 1-2-chloroethyl-, hydrochloride, 2-piperidinoethyl chloride hydrochloride, piperidinoethyl chloride, hydrochloride, beta-chloroethylpiperidine hydrochloride, 1-2-chloroethyl piperidinium chloride, n-2-chloroethyl piperidine hydrochloride
PubChem CID 74826
IUPAC Name 1-(2-chloroethyl)piperidine;hydrochloride
SMILES C1CCN(CC1)CCCl.Cl

Specifications

Melting Point 232°C
Color White-Yellow
Quantity 500 g
UN Number 2928
Formula Weight 184.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2-Chloroethyl)piperidine Hydrochloride
EINECSNumber (9)-1125
RTECSNumber TM6482500
TSCA No