missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Chloroethyl)piperidine hydrochloride, 98%

Catalog No. p-7026775
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2298518 50 g
AAB2298530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2298518 Supplier Thermo Scientific Chemicals Supplier No. B2298518
Only null left

CAS: 2008-75-5 | C7H15Cl2N | 184.104 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2008-75-5
Molecular Formula C7H15Cl2N
Molecular Weight (g/mol) 184.104
MDL Number MFCD00012837
InChI Key VFLQQZCRHPIGJU-UHFFFAOYSA-N
Synonym 1-2-chloroethyl piperidine hydrochloride, 2-piperidinoethylchloride hydrochloride, 2-chloroethylpiperidine hydrochloride, n-chloroethyl piperidine hydrochloride, piperidine, 1-2-chloroethyl-, hydrochloride, 2-piperidinoethyl chloride hydrochloride, piperidinoethyl chloride, hydrochloride, beta-chloroethylpiperidine hydrochloride, 1-2-chloroethyl piperidinium chloride, n-2-chloroethyl piperidine hydrochloride
PubChem CID 74826
IUPAC Name 1-(2-chloroethyl)piperidine;hydrochloride
SMILES C1CCN(CC1)CCCl.Cl

Specifications

Melting Point 229°C to 235°C
Quantity 50 g
UN Number UN2928
Beilstein 3910268
Sensitivity Hygroscopic
Formula Weight 184.11
Percent Purity 98%
Chemical Name or Material 1-(2-Chloroethyl)piperidine hydrochloride
Hazard Category H300-H314-H335-H341
Hazard Statement GHS H Statement
H300-H314-H318-H341
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing genetic defects.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.
EINECSNumber 217-920-5
RTECSNumber TM6482500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only