missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Chloroethyl)piperidine hydrochloride, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 109215000

Quantity 500g
Packaging Plastic bottle

 View more versions of this product

Catalog No. AC109215000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2008-75-5
184.11
VFLQQZCRHPIGJU-UHFFFAOYSA-N
74826
C1CCN(CC1)CCCl.Cl
C7H14ClN·HCl
MFCD00012837
1-2-chloroethyl piperidine hydrochloride, 2-piperidinoethylchloride hydrochloride, 2-chloroethylpiperidine hydrochloride, n-chloroethyl piperidine hydrochloride, piperidine, 1-2-chloroethyl-, hydrochloride, 2-piperidinoethyl chloride hydrochloride, piperidinoethyl chloride, hydrochloride, beta-chloroethylpiperidine hydrochloride, 1-2-chloroethyl piperidinium chloride, n-2-chloroethyl piperidine hydrochloride
1-(2-chloroethyl)piperidine;hydrochloride

Specifications

1-(2-Chloroethyl)piperidine hydrochloride
2008-75-5
100.0
Authentic
Plastic bottle
500g
20, 17
Solubility in water: soluble. Other solubilities: soluble in hot ethanol and hot acetone,fairly soluble in cold ethanol,sparingly soluble in cold acetone,insoluble in benzene and toluene
C1CCN(CC1)CCCl.Cl
184.11
184.11
98%
97.5
1% max. (105°C, 3 hrs)
98%
C7H14ClN·HCl
MFCD00012837
1-2-chloroethyl piperidine hydrochloride, 2-piperidinoethylchloride hydrochloride, 2-chloroethylpiperidine hydrochloride, n-chloroethyl piperidine hydrochloride, piperidine, 1-2-chloroethyl-, hydrochloride, 2-piperidinoethyl chloride hydrochloride, piperidinoethyl chloride, hydrochloride, beta-chloroethylpiperidine hydrochloride, 1-2-chloroethyl piperidinium chloride, n-2-chloroethyl piperidine hydrochloride
VFLQQZCRHPIGJU-UHFFFAOYSA-N
1-(2-chloroethyl)piperidine;hydrochloride
74826
98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if swallowed.
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wash face,ha

GHS Signal Word: Danger

EINECSNumber : 217-920-5

RTECSNumber : TM6482500

TSCA : TSCA

SDS
Documents

Documents