missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Chloroethyl)-2-pyrrolidone, 95%

Catalog No. AC453760050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453760050 5 g
AC453760010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC453760050 Supplier Thermo Scientific Chemicals Supplier No. 453760050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51333-90-5 | C6H10ClNO | 147.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51333-90-5
Molecular Formula C6H10ClNO
Molecular Weight (g/mol) 147.60
MDL Number MFCD00169064
InChI Key CWLYHDWHNFLUEI-UHFFFAOYSA-N
Synonym 1-2-chloroethyl pyrrolidin-2-one, 1-2-chloro-ethyl-pyrrolidin-2-one, 1-2-chloroethyl-2-pyrrolidinone, 2-pyrrolidinone, 1-2-chloroethyl, n-2-chloroethyl-2-pyrrolidone, 1-2-chloroethyl-2-pyrrolidone, n-beta-chloroethyl-pyrrolidinone, 1-2-chloroethyl-pyrrolidin-2-one, n-beta-chloroethyl-2-pyrrolidinone
PubChem CID 3156650
IUPAC Name 1-(2-chloroethyl)pyrrolidin-2-one
SMILES ClCCN1CCCC1=O

Specifications

Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Quantity 5 g
Formula Weight 147.61
Percent Purity 95%
Chemical Name or Material 1-(2-Chloroethyl)-2-pyrrolidone
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only