missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Chloroethyl)-1H-hexahydroazepine Hydrochloride 98.0+%, TCI America™

Catalog No. C23795G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23795G 5 g
C237925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23795G Supplier TCI America Supplier No. C23795G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26487-67-2
Molecular Formula C8H17Cl2N
Molecular Weight (g/mol) 198.131
MDL Number MFCD00012842
InChI Key ZQDSOUPBYJIPNM-UHFFFAOYSA-N
Synonym 1-2-chloroethyl azepane hydrochloride, 2-hexamethyleneimino ethyl chloride hydrochloride, 1h-azepine, hydrochloride, 2-chloroethylhexamethylenemine hydrochloride, 1-2-chloroethyl hexamethyleneimine hydrochloride, 2-chloroethyl hexamethylene imine hydrochloride, 2-chloroethyl azaperhydroepine, chloride, pubchem21389, 1-2-chloroethyl azepane hcl, wln: t7ntj a2g &gh
PubChem CID 24188283
IUPAC Name 1-(2-chloroethyl)azepane;hydrochloride
SMILES C1CCCN(CC1)CCCl.Cl

Specifications

Melting Point 208°C
Color White-Yellow
Quantity 5 g
Formula Weight 198.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2-Chloroethyl)-1H-hexahydroazepine Hydrochloride
RTECSNumber CM3185000
TSCA No