missing translation for 'onlineSavingsMsg'
Learn More

1,2-Butylene Oxide 99.0+%, TCI America™

Catalog No. B0719500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0719500ML 500 mL
B071925ML 25 mL
B0719100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0719500ML Supplier TCI America Supplier No. B0719500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 106-88-7
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.107
MDL Number MFCD00005153
InChI Key RBACIKXCRWGCBB-UHFFFAOYSA-N
Synonym 1,2-epoxybutane, 1,2-butylene oxide, oxirane, ethyl, ethyloxirane, 1,2-butene oxide, ethylethylene oxide, 1-butene oxide, oxirane, 2-ethyl, butane, epoxy, 1-butylene oxide
PubChem CID 7834
ChEBI CHEBI:82326
IUPAC Name 2-ethyloxirane
SMILES CCC1CO1

Specifications

Color Colorless
Boiling Point 63°C
Quantity 500 mL
UN Number 3022
Formula Weight 72.11
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Butylene Oxide
EINECSNumber (2)-0229
RTECSNumber EK3675000
TSCA Yes